accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHBr

CAS RN 25574-11-2

IUPAC Name: 3-(4-bromophenyl)propan-1-ol
Synonyms: 3-(4-bromophenyl)propan-1-ol
Molecular Weight (g/mol): 215.09
Molecular Formula: C9H11BrO
InChi Key: WODKXGCVVOOEIJ-UHFFFAOYSA-N
SMILES: OCCCC1=CC=C(Br)C=C1
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)