Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(4-Bromophenyl)-1-propanol 98.0+%, TCI America™
SDP

Catalog No. B46575G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B46575G 5 g
B46571G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B46575G Supplier TCI America Supplier No. B46575G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 25574-11-2
Molecular Formula C9H11BrO
Molecular Weight (g/mol) 215.09
MDL Number MFCD09028724
InChI Key WODKXGCVVOOEIJ-UHFFFAOYSA-N
Synonym 3-4-bromophenyl propan-1-ol, 4-bromobenzenepropanol, 3-4-bromo-phenyl-propan-1-ol, benzenepropanol, 4-bromo, 3-4-bromophenyl-1-propanol, pubchem3984, 3-4-bromophenyl propanol, 3-4-bromophenyl-propanol, amth183, 3-4-bromophenyl propyl alcohol
PubChem CID 10560614
IUPAC Name 3-(4-bromophenyl)propan-1-ol
SMILES C1=CC(=CC=C1CCCO)Br

Specifications

Color Yellow
Boiling Point 162°C
Quantity 5 g
Formula Weight 215.09
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-(4-Bromophenyl)-1-propanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.