accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOOCH2

CAS RN 25584-83-2

IUPAC Name: 3-hydroxypropyl prop-2-enoate
Synonyms: hydroxypropyl acrylate
Molecular Weight (g/mol): 130.14
Molecular Formula: C6H10O3
InChi Key: QZPSOSOOLFHYRR-UHFFFAOYSA-N
SMILES: OCCCOC(=O)C=C
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)