Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Hydroxypropyl acrylate, mixture of isomers

Catalog No. 3709321L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 L
18 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
37-093-21L 1 L
37-093-218L 18 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 37-093-21L Supplier Sigma Aldrich Supplier No. 3709321L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Hydroxypropyl acrylates may be used for the synthesis of a number of acrylates. The presence of both vinyl and hydroxyl allows copolymerization of the acrylate with vinyl monomers as well as with di- or polyisocyanates, anhydrides, or epoxides.

Specifications

CAS 25584-83-2
Density 1.044 g/mL (at 25°C)
Boiling Point 77°C (5 mmHg)
Molecular Formula C6H10O3
Refractive Index n20/D 1.445 (literature)
Linear Formula H2C=CHCO2C3H6OH
MDL Number MFCD04113589
Quantity 1 L
Molecular Weight (g/mol) 130.14
Percent Purity 95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.