accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OSOCH3

CAS RN 255901-50-9

IUPAC Name: 3,3-dimethyl-2H,3H,4H-thieno[3,4-b][1,4]dioxepine
Synonyms: 3,3-dimethyl-2H,4H-thieno[3,4-b][1,4]dioxepine
Molecular Weight (g/mol): 184.25
Molecular Formula: C9H12O2S
InChi Key: PUEUIEYRIVFGLS-UHFFFAOYSA-N
SMILES: CC1(C)COC2=CSC=C2OC1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)