accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 256-96-2

IUPAC Name: 2-azatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,9,11,13-heptaene
Synonyms: dibenzazepine 5h-dibenz(b,f)azepine 2,3,6,7-dibenzazepine 5H-dibenzo(b,f)azepine 5H-dibenz(b,f)azepine 10-(13)c-labeled iminostilbene 10-(13)C-labeled iminostilbene iminostilbene dibenz(b,f)azepine
Molecular Weight (g/mol): 193.25
Molecular Formula: C14H11N
InChi Key: LCGTWRLJTMHIQZ-UHFFFAOYSA-N
SMILES: N1C2=CC=CC=C2C=CC2=CC=CC=C12
Molecular Weight (g/mol) 
  • (1)
  • (5)
  • (2)
Percent Purity 
  • (2)
  • (6)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (2)
  • (5)
  • (1)
  • (5)
  • (2)
  • (2)
  • (6)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)