accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH3CH2CH3OOCH3H2C

CAS RN 25608-33-7

IUPAC Name: butyl 2-methylprop-2-enoate; methyl 2-methylprop-2-enoate
Synonyms: butyl methacrylate; methyl methacrylate
Molecular Weight (g/mol): 242.32
Molecular Formula: C13H22O4
InChi Key: WHLPIOPUASGRQN-UHFFFAOYSA-N
SMILES: COC(=O)C(C)=C.CCCCOC(=O)C(C)=C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results