Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Poly(butyl methacrylate-co-methyl methacrylate)

Catalog No. 474037250G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47-403-7250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 47-403-7250G Supplier Sigma Aldrich Supplier No. 474037250G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Poly(butyl methacrylate-co-methyl methacrylate) (P(BMA-co-MMA) ) may be used to synthesize porous interpenetrating polymer network (IPN) with low-density polymers. These IPN structures are applicable as membranes for separation of liquid mixtures due to their high mechanical strength and selectivity. P(BMA-co-MMA) particles may be used to encapsulate single non-aggregated semiconducting quantum dots (QDs) for size enlargement.These hybrid organic/inorganic nanostructures may be used as single photon sources.

Specifications

CAS 25608-33-7
Density 1.08 g/mL (at 25°C)
Linear Formula [CH2C(CH3)[CO2(CH2)3 CH3]]X[CH2C(CH3)(CO2CH3)]Y
MDL Number MFCD00145620
Quantity 250 g
Molecular Weight (g/mol) 150000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.