accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2561-85-5

IUPAC Name: 3-dodecyloxolane-2,5-dione
Synonyms: 3-dodecyloxolane-2,5-dione
Molecular Weight (g/mol): 268.4
Molecular Formula: C16H28O3
InChi Key: YAXXOCZAXKLLCV-UHFFFAOYNA-N
SMILES: CCCCCCCCCCCCC1CC(=O)OC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results