Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dodecylsuccinic Anhydride 95.0+%, TCI America™
SDP

Catalog No. D097725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D097725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D097725G Supplier TCI America Supplier No. D097725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2561-85-5
Molecular Formula C16H28O3
Molecular Weight (g/mol) 268.40
MDL Number MFCD00022610
InChI Key YAXXOCZAXKLLCV-UHFFFAOYNA-N
PubChem CID 98107
IUPAC Name 3-dodecyloxolane-2,5-dione
SMILES CCCCCCCCCCCCC1CC(=O)OC1=O

Specifications

Melting Point 73°C
Color White
Quantity 25 g
Physical Form Crystal-Powder at 20°C
Formula Weight 268.40
Percent Purity ≥95.0% (GC,T)
Chemical Name or Material Dodecylsuccinic Anhydride
EINECSNumber (2)-0851
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.