accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OCH3HO

CAS RN 2563-07-7

IUPAC Name: 2-ethoxy-4-methylphenol
Synonyms: ethoxy-P-cresol
Molecular Weight (g/mol): 152.19
Molecular Formula: C9H12O2
InChi Key: ZWQBCCVEOIYNDL-UHFFFAOYSA-N
SMILES: CCOC1=CC(C)=CC=C1O
Boiling Point 
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results