CAS RN 2565-58-4
IUPAC Name:
(chloromethyl)phosphonic acid
Synonyms:
chloromethylphosphonic acid
Molecular Weight (g/mol):
130.46
Molecular Formula:
CH4ClO3P
InChi Key:
MOFCYHDQWIZKMY-UHFFFAOYSA-N
SMILES:
OP(O)(=O)CCl
1
–
1
of
1
results
1
Promotions Available
Chloromethylphosphonic acid, 98%, Thermo Scientific Chemicals
Chloromethylphosphonic acid, 98%, Thermo Scientific Chemicals
CAS: 2565-58-4 Molecular Formula: CH4ClO3P Molecular Weight (g/mol): 130.46 MDL Number: MFCD00013955 InChI Key: MOFCYHDQWIZKMY-UHFFFAOYSA-N Synonym: chloromethyl phosphonic acid,phosphonic acid, chloromethyl,chloromethanephosphonic acid,phosphonic acid, p-chloromethyl,monochloromethyl phosphonic acid,chloromethylphosphite,phosphonomethyl chloride,chloromethyl-phosphonicaci,phosphonic acid,p-chloromethyl,chloromethyl phosphonic acid, 98 % PubChem CID: 75723 IUPAC Name: chloromethylphosphonic acid SMILES: OP(O)(=O)CCl