Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloromethylphosphonic acid, 98%, Thermo Scientific Chemicals

Catalog No. AC451600050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC451600050 5 g Glass Bottle
AC451600010 1 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC451600050 Supplier Thermo Scientific Chemicals Supplier No. 451600050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2565-58-4
Molecular Formula CH4ClO3P
Molecular Weight (g/mol) 130.46
MDL Number MFCD00013955
InChI Key MOFCYHDQWIZKMY-UHFFFAOYSA-N
Synonym chloromethyl phosphonic acid, phosphonic acid, chloromethyl, chloromethanephosphonic acid, phosphonic acid, p-chloromethyl, monochloromethyl phosphonic acid, chloromethylphosphite, phosphonomethyl chloride, chloromethyl-phosphonicaci, phosphonic acid,p-chloromethyl, chloromethyl phosphonic acid, 98 %
PubChem CID 75723
IUPAC Name chloromethylphosphonic acid
SMILES OP(O)(=O)CCl

Specifications

Melting Point 88.0°C to 92.0°C
Assay Percent Range 98%
Packaging Glass Bottle
Quantity 5 g
Formula Weight 130.47
Percent Purity 98%
Chemical Name or Material Chloromethylphosphonic Acid
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF SWALLOWED
EINECSNumber 219-895-6

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.