accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 256506-99-7

IUPAC Name: ethyl 2-amino-4-(furan-2-yl)thiophene-3-carboxylate
Synonyms: ethyl 2-amino-4-(furan-2-yl)thiophene-3-carboxylate
Molecular Weight (g/mol): 237.27
Molecular Formula: C11H11NO3S
InChi Key: JMLBJOYPNPOBKV-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(N)SC=C1C1=CC=CO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results