CAS RN 2566-22-5
CAS RN 2566-22-5
IUPAC Name:
3-phenyl-2-(phenylformamido)propanoic acid
Synonyms:
3-phenyl-2-(phenylformamido)propanoic acid
Molecular Weight (g/mol):
269.3
Molecular Formula:
C16H15NO3
InChi Key:
NPKISZUVEBESJI-UHFFFAOYNA-N
SMILES:
OC(=O)C(CC1=CC=CC=C1)NC(=O)C1=CC=CC=C1
1
–
1
of
1
results
1
Advanced Chem Tech Bz-Phe-OH, Advanced ChemTech
CAS: 2566-22-5 Molecular Formula: C16H15NO3 Molecular Weight (g/mol): 269.30 InChI Key: NPKISZUVEBESJI-UHFFFAOYNA-N IUPAC Name: 3-phenyl-2-(phenylformamido)propanoic acid SMILES: OC(=O)C(CC1=CC=CC=C1)NC(=O)C1=CC=CC=C1