accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2566-30-5

OHONH(S)CH3

CAS RN 2566-30-5

IUPAC Name: (2S)-2-(methylamino)-3-phenylpropanoic acid
Synonyms: N-me-phe|N-me-phe-OH|N-me-L-phe|N-me-L-phe-OH
Molecular Weight (g/mol): 179.22
Molecular Formula: C10H13NO2
InChi Key: SCIFESDRCALIIM-VIFPVBQESA-N
SMILES: CN[C@@H](CC1=CC=CC=C1)C(O)=O
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)