Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-MePhe-OH.HCl, Advanced ChemTech

Catalog No. 01338455
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-338-455 1 g
01-338-454 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-338-455 Supplier Advanced Chem Tech Supplier No. UM87451G
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2566-30-5
Molecular Formula C10H13NO2
Molecular Weight (g/mol) 179.22
MDL Number MFCD00037757
InChI Key SCIFESDRCALIIM-VIFPVBQESA-N
IUPAC Name (2S)-2-(methylamino)-3-phenylpropanoic acid
SMILES CN[C@@H](CC1=CC=CC=C1)C(O)=O

Specifications

Color White/Off-White
Quantity 1 g
Grade Biochemical
Chemical Name or Material N-MePhe-OH.HCl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.