accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 25676-75-9

IUPAC Name: 4-bromo-1-methyl-1H-imidazole
Synonyms: 4-bromo-1-methylimidazole
Molecular Weight (g/mol): 161
Molecular Formula: C4H5BrN2
InChi Key: IOTSLMMLLXTNNH-UHFFFAOYSA-N
SMILES: CN1C=NC(Br)=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)