accessibility menu, dialog, popup

UserName

Viewing profile as user:
NN

CAS RN 25677-69-4

IUPAC Name: 2,9-diphenyl-1,10-phenanthroline
Synonyms: 2,9-diphenyl-1,10-phenanthroline
Molecular Weight (g/mol): 332.41
Molecular Formula: C24H16N2
InChi Key: HVCVRVKEIKXBIF-UHFFFAOYSA-N
SMILES: C1=CC=C(C=C1)C1=CC=C2C=CC3=CC=C(N=C3C2=N1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results