accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SH

CAS RN 25697-56-7

IUPAC Name: (3-methylphenyl)methanethiol
Synonyms: (3-methylphenyl)methanethiol
Molecular Weight (g/mol): 138.23
Molecular Formula: C8H10S
InChi Key: PNAZUQUHTIOEHF-UHFFFAOYSA-N
SMILES: CC1=CC(CS)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results