Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(3-Methylphenyl)methanethiol, 95%, Thermo Scientific™

Catalog No. TL00469EB
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
TL00469EB 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. TL00469EB Supplier Thermo Scientific Chemicals Supplier No. TL00469EB
Add to Cart

Chemical Identifiers

CAS 25697-56-7
Molecular Formula C8H10S
Molecular Weight (g/mol) 138.228
MDL Number MFCD00022075
InChI Key PNAZUQUHTIOEHF-UHFFFAOYSA-N
PubChem CID 2801207
IUPAC Name (3-methylphenyl)methanethiol
SMILES CC1=CC(=CC=C1)CS

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Linear Formula CH3C6H4CH2SH
Quantity 25 g
Synonym 3-methylphenyl methanethiol, 3-methylbenzyl mercaptan, m-methylbenzyl mercaptan, 3-methylbenzenemethanethiol, m-methylbenzylmercaptan, acmc-1cq7k, 3-methyl-benzenemethanethiol, benzenemethanethiol,3-methyl, benzenemethanethiol, 3-methyl, 3-methylphenyl methane-1-thiol
Formula Weight 138.23
Percent Purity 95%
Chemical Name or Material (3-Methylphenyl)methanethiol
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.