accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 25713-60-4

IUPAC Name: tris(2,4,6-tribromophenoxy)-1,3,5-triazine
Synonyms: tris(2,4,6-tribromophenoxy)-1,3,5-triazine
Molecular Weight (g/mol): 1067.43
Molecular Formula: C21H6Br9N3O3
InChi Key: BDFBPPCACYFGFA-UHFFFAOYSA-N
SMILES: BrC1=CC(Br)=C(OC2=NC(OC3=C(Br)C=C(Br)C=C3Br)=NC(OC3=C(Br)C=C(Br)C=C3Br)=N2)C(Br)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results