Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,6-Tris(2,4,6-tribromophenoxy)-1,3,5-triazine 98.0+%, TCI America™
SDP

Catalog No. T28715G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T28715G 5 g
T287125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T28715G Supplier TCI America Supplier No. T28715G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 25713-60-4
Molecular Formula C21H6Br9N3O3
Molecular Weight (g/mol) 1067.433
MDL Number MFCD03092948
InChI Key BDFBPPCACYFGFA-UHFFFAOYSA-N
Synonym Cyanuric Acid Tris(2,4,6-tribromophenyl) Ester, Tris(2,4,6-tribromophenyl) Cyanurate
PubChem CID 91820
IUPAC Name 2,4,6-tris(2,4,6-tribromophenoxy)-1,3,5-triazine
SMILES C1=C(C=C(C(=C1Br)OC2=NC(=NC(=N2)OC3=C(C=C(C=C3Br)Br)Br)OC4=C(C=C(C=C4Br)Br)Br)Br)Br

Specifications

Melting Point 230°C
Color White
Quantity 5 g
Formula Weight 1067.43
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2,4,6-Tris(2,4,6-tribromophenoxy)-1,3,5-triazine
EINECSNumber (5)-6312
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.