accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 257610-49-4

IUPAC Name: (1-fluoroethenyl)(methyl)diphenylsilane
Synonyms: (1-fluoroethenyl)(methyl)diphenylsilane
Molecular Weight (g/mol): 242.37
Molecular Formula: C15H15FSi
InChi Key: IBMIUWHXFMPYLJ-UHFFFAOYSA-N
SMILES: C[Si](C(F)=C)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results