Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1-Fluorovinyl)methyldiphenylsilane 96.0+%, TCI America™
SDP

Catalog No. F05185G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F05185G 5 g
F05181G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F05185G Supplier TCI America Supplier No. F05185G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 257610-49-4
Molecular Formula C15H15FSi
Molecular Weight (g/mol) 242.37
MDL Number MFCD08276379
InChI Key IBMIUWHXFMPYLJ-UHFFFAOYSA-N
Synonym 1-Fluoro-1-(methyldiphenylsilyl)ethylene
PubChem CID 10922738
IUPAC Name (1-fluoroethenyl)(methyl)diphenylsilane
SMILES C[Si](C(F)=C)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color Yellow
Quantity 5 g
Formula Weight 242.37
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material (1-Fluorovinyl)methyldiphenylsilane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.