accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 25774-02-1

IUPAC Name: 3-(benzyloxy)-3-oxo-2-phenylpropanoic acid
Synonyms: 3-(benzyloxy)-3-oxo-2-phenylpropanoic acid
Molecular Weight (g/mol): 270.28
Molecular Formula: C16H14O4
InChi Key: QSBAHMROFICXDC-UHFFFAOYNA-N
SMILES: OC(=O)C(C(=O)OCC1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results