Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Monobenzyl Phenylmalonate 97.0+%, TCI America™
SDP

Catalog No. P13415G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P13415G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P13415G Supplier TCI America Supplier No. P13415G

Chemical Identifiers

CAS 25774-02-1
Molecular Formula C16H14O4
Molecular Weight (g/mol) 270.284
MDL Number MFCD00051723
InChI Key QSBAHMROFICXDC-UHFFFAOYSA-N
Synonym Phenylmalonic Acid Monobenzyl Ester
PubChem CID 117278
IUPAC Name 3-oxo-2-phenyl-3-phenylmethoxypropanoic acid
SMILES C1=CC=C(C=C1)COC(=O)C(C2=CC=CC=C2)C(=O)O

Specifications

Melting Point 66°C
Color White-Yellow
Quantity 5 g
Formula Weight 270.28
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Monobenzyl Phenylmalonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.