accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHHO

CAS RN 25779-13-9

IUPAC Name: 1-phenylethane-1,2-diol
Synonyms: 1,2-ethanediol, 1-phenyl-
Molecular Weight (g/mol): 138.17
Molecular Formula: C8H10O2
InChi Key: PWMWNFMRSKOCEY-UHFFFAOYNA-N
SMILES: OCC(O)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (4)
Quantity 
  • (3)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (4)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)