Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(+)-1-Phenylethane-1,2-diol 98.0+%, TCI America™
SDP

Catalog No. P11515G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P11515G 5 g
P11511G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P11515G Supplier TCI America Supplier No. P11515G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 25779-13-9
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.166
MDL Number MFCD00066256
InChI Key PWMWNFMRSKOCEY-MRVPVSSYSA-N
Synonym s-+-1-phenyl-1,2-ethanediol, s-1-phenylethane-1,2-diol, 1s-1-phenylethane-1,2-diol, s-+-1-phenylethane-1,2-diol, s-1-phenyl-1,2-ethanediol, unii-8ib2t1ft6b, 8ib2t1ft6b, 1,2-ethanediol, 1-phenyl-, 1s, styrene glycol, +, r---phenylethylene glycol
PubChem CID 643312
IUPAC Name (1S)-1-phenylethane-1,2-diol
SMILES C1=CC=C(C=C1)C(CO)O

Specifications

Melting Point 68°C
Color White
Boiling Point 82°C
Quantity 5 g
Formula Weight 138.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (S)-(+)-1-Phenylethane-1,2-diol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.