accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2578-33-8

OOOOHNH2

CAS RN 2578-33-8

IUPAC Name: 2-amino-5-(benzyloxy)-5-oxopentanoic acid
Synonyms: 2-amino-5-(benzyloxy)-5-oxopentanoic acid
Molecular Weight (g/mol): 237.26
Molecular Formula: C12H15NO4
InChi Key: BGGHCRNCRWQABU-UHFFFAOYNA-N
SMILES: NC(CCC(=O)OCC1=CC=CC=C1)C(O)=O
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)