accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONONClNOO

CAS RN 2578-45-2

IUPAC Name: 2-chloro-3,5-dinitropyridine
Synonyms: 2-chloro-3,5-dinitropyridine
Molecular Weight (g/mol): 203.54
Molecular Formula: C5H2ClN3O4
InChi Key: QLHVJBXAQWPEDI-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CN=C(Cl)C(=C1)[N+]([O-])=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)