Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-3,5-dinitropyridine, 98+%

Catalog No. AC208190050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC208190050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC208190050 Supplier Thermo Scientific Chemicals Supplier No. 208190050

missing translation for 'validMainframeMsg'

CAS: 2578-45-2 | C5H2ClN3O4 | 203.54 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2578-45-2
Molecular Formula C5H2ClN3O4
Molecular Weight (g/mol) 203.54
MDL Number MFCD00006233
InChI Key QLHVJBXAQWPEDI-UHFFFAOYSA-N
Synonym pyridine, 2-chloro-3,5-dinitro, 2-chloro-3,5-dinitro-pyridine, pubchem6103, acmc-209gmh, 2-chloro3,5-dinitropyridine, 2-chloro-3,5-dinitropyridine
PubChem CID 75738
IUPAC Name 2-chloro-3,5-dinitropyridine
SMILES C1=C(C=NC(=C1[N+](=O)[O-])Cl)[N+](=O)[O-]

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 63.0°C to 66.0°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula (O2H)(C5H2N)Cl
Quantity 5 g
Beilstein 20,III,2536
Formula Weight 203.54
Percent Purity 98+%
Physical Form Powder
Chemical Name or Material 2-Chloro-3,5-dinitropyridine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Fatal if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
Wear protective gloves/protective clothing
EINECSNumber 219-937-3

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.