accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH3(S)(R)NH3

CAS RN 2579-20-6

IUPAC Name: [(1R,3S)-3-(azaniumylmethyl)cyclohexyl]methanaminium
Synonyms: [(1R,3S)-3-(ammoniomethyl)cyclohexyl]methanaminium
Molecular Weight (g/mol): 144.26
Molecular Formula: C8H20N2
InChi Key: QLBRROYTTDFLDX-OCAPTIKFSA-P
SMILES: [NH3+]C[C@H]1CCC[C@@H](C[NH3+])C1
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (1)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (4)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)
  • (3)