Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1,3-Cyclohexanebis(methylamine), mixture of isomers

Catalog No. 180467100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-046-7100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-046-7100G Supplier Sigma Aldrich Supplier No. 180467100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.1,3-Cyclohexanebis(methylamine), mixture of isomers (1,3-BAC), is used as a cross-linking agent that can be used in the formation of fiber-based membranes for the separation of CO2. It can also be used in the synthesis of open-framework bimetallic phosphites for potential usage in molecular sieves and ion exchangers.

Specifications

CAS 2579-20-6
Density 0.945 g/mL (at 25°C)
Molecular Formula C8H18N2
Refractive Index n20/D 1.493 (literature)
Linear Formula C6H10(CH2NH2)2
Quantity 100 g
MDL Number MFCD00001522
Synonym 1,3-Bis(aminomethyl)cyclohexane
Molecular Weight (g/mol) 142.24
Percent Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.