accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClCH3NCH3CNH3CCH3HN

CAS RN 2580-56-5

IUPAC Name: bis[4-(dimethylamino)phenyl][4-(phenylamino)naphthalen-1-yl]methylium chloride
Synonyms: bis[4-(dimethylamino)phenyl][4-(phenylamino)naphthalen-1-yl]methylium chloride
Molecular Weight (g/mol): 506.09
Molecular Formula: C33H32ClN3
InChi Key: QHHVGBIWIQLWOJ-UHFFFAOYSA-M
SMILES: [Cl-].CN(C)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(C)C)C1=C2C=CC=CC2=C(NC2=CC=CC=C2)C=C1
Color 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Grade 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)