Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Victoria blue B, MP Biomedicals™

Catalog No. ICN15829125 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15829125 25 g
ICN15829180 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15829125 Supplier MP Biomedicals, Inc Supplier No. 0215829125
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2580-56-5
Molecular Formula C33H32ClN3
Molecular Weight (g/mol) 506.09
InChI Key LLWJPGAKXJBKKA-UHFFFAOYSA-N
Synonym Basic Blue B, Basic Blue 26
PubChem CID 17407
ChEBI CHEBI:87651
IUPAC Name [4-[bis[4-(dimethylamino)phenyl]methylidene]naphthalen-1-ylidene]-phenylazanium;chloride
SMILES CN(C)C1=CC=C(C=C1)C(=C2C=CC(=[NH+]C3=CC=CC=C3)C4=CC=CC=C24)C5=CC=C(C=C5)N(C)C.[Cl-]

Specifications

Color Blue to Brown
Quantity 25 g
Physical Form Powder
Formula Weight 506.1
Grade Not Available
Chemical Name or Material Victoria blue B
EINECSNumber 219-943-6
Recommended Storage Store at room temperature.
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.