accessibility menu, dialog, popup

UserName

Viewing profile as user:
∗N∗OMen

CAS RN 25805-17-8

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): —
Molecular Formula: (C5H9NO)n
InChi Key: —
SMILES: CCC(=O)N(-*)CC-*
Molecular Weight (g/mol) 
  • (2)
  • (1)
  • (9)
Quantity 
  • (5)
  • (3)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (9)
  • (2)
  • (1)
  • (9)
  • (5)
  • (3)
  • (4)