Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Poly(2-ethyl-2-oxazoline)

Catalog No. 372846500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
37-284-6500G 500 g
37-284-6100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 37-284-6500G Supplier Sigma Aldrich Supplier No. 372846500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

This Poly(2-ethyl-2-oxazoline) polymer is amorphous and water soluble with good temperature stability. Jordan and coworkers showed biocompatibility, no accumulation in tissue, and rapid clearance from the bloodstream. End-group modified poly(2-ethyl-2-oxazoline)s have been conjugated to peptides, and were shown as versatile alternatives to poly(ethylene glycol) (PEG) for both protein and small drug conjugation.Potential substitute for poly(vinyl alcohol) and poly(vinyl pyrrolidone). Adhesion promoter in coatings. Heat sealing and remoistenable hot-melt adhesive

Specifications

CAS 25805-17-8
Density 1.14 g/mL (at 25°C)
Refractive Index n20/D 1.52
Linear Formula [-N(COC2H5)CH2CH2-]n
Quantity 500 g
MDL Number MFCD00134377
Synonym PEOX; POx; PetOx; poly 2-ethyloxazoline; polyethyloxazoline
Molecular Weight (g/mol) 50000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.