accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 258273-75-5

IUPAC Name: butyltrimethylazanium; trifluoro[(trifluoromethanesulfonylazanidyl)sulfonyl]methane
Synonyms: bistriflylimide anion; butyltrimethylazanium
Molecular Weight (g/mol): 396.36
Molecular Formula: C9H18F6N2O4S2
InChi Key: XSGKJXQWZSFJEJ-UHFFFAOYSA-N
SMILES: CCCC[N+](C)(C)C.FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results