Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Butyltrimethylammonium Bis(trifluoromethanesulfonyl)imide 98.0+%, TCI America™
SDP

Catalog No. B32335G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B32335G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B32335G Supplier TCI America Supplier No. B32335G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 258273-75-5
Molecular Formula C9H18F6N2O4S2
Molecular Weight (g/mol) 396.363
MDL Number MFCD12761449
InChI Key XSGKJXQWZSFJEJ-UHFFFAOYSA-N
PubChem CID 12991988
IUPAC Name bis(trifluoromethylsulfonyl)azanide;butyl(trimethyl)azanium
SMILES CCCC[N+](C)(C)C.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F

Specifications

Melting Point 17°C
Quantity 5 g
Formula Weight 396.36
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material Butyltrimethylammonium Bis(trifluoromethanesulfonyl)imide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.