accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3NH2

CAS RN 25834-93-9

IUPAC Name: 4-(butan-2-yl)cyclohexan-1-amine
Synonyms: 4-(sec-butyl)cyclohexan-1-amine
Molecular Weight (g/mol): 155.29
Molecular Formula: C10H21N
InChi Key: PVQLHUCSNDRLDP-UHFFFAOYNA-N
SMILES: CCC(C)C1CCC(N)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results