Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-sec-Butylcyclohexylamine (cis- and trans- mixture) 98.0+%, TCI America™
SDP

Catalog No. B40045ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B40045ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B40045ML Supplier TCI America Supplier No. B40045ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 25834-93-9
Molecular Formula C10H21N
Molecular Weight (g/mol) 155.285
InChI Key PVQLHUCSNDRLDP-UHFFFAOYSA-N
Synonym 4-Amino-1-sec-butylcyclohexane
PubChem CID 22561486
IUPAC Name 4-butan-2-ylcyclohexan-1-amine
SMILES CCC(C)C1CCC(CC1)N

Specifications

Color Colorless
Boiling Point 105°C
Quantity 5 mL
UN Number 2735
Formula Weight 155.29
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-sec-Butylcyclohexylamine (cis- and trans- mixture)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.