accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 25852-47-5

IUPAC Name: 2-[(2-methylprop-2-enoyl)oxy]ethyl 2-methylprop-2-enoate
Synonyms: glycol dimethacrylate ethylene dimethacrylate ethanediol dimethacrylate glycol hema-methacrylate ethylene glycol dimethacrylate 1,2-ethanediol dimethacrylate diglycol dimethacrylate ethyldiol metacrylate ethyldiol methacrylate ethylenedimethyacrylate
Molecular Weight (g/mol): 198.22
Molecular Formula: C10H14O4
InChi Key: STVZJERGLQHEKB-UHFFFAOYSA-N
SMILES: CC(=C)C(=O)OCCOC(=O)C(C)=C
Quantity 
  • (2)
  • (8)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (10)
Molecular Weight (g/mol) 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (10)
  • (2)
  • (5)
  • (2)
  • (8)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (10)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)