Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Poly(ethylene glycol) dimethacrylate

Catalog No. 9068591G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-685-91G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 90-685-91G Supplier Sigma Aldrich Supplier No. 9068591G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

High purity PEG dimethacrylates are commonly used in PEG-based hydrogels and crosslinking hydrophilic polymers. It is good for tissue engineering and 3D printing applications.

Specifications

CAS 25852-47-5
Boiling Point >200°C (2 mmHg)
Linear Formula C3H5C(O)(OCH2CH2)nOC(O)C3H5
MDL Number MFCD00081877
Quantity 1 g
Synonym PEG bismethacrylate; PEG hydrogel; PEG dimethacrylate
Molecular Weight (g/mol) 1000
Percent Purity 99% (NMR)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.