accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COSeOOCH3

CAS RN 25862-14-0

IUPAC Name: 1-methoxy-4-(4-methoxybenzeneseleninyl)benzene
Synonyms: 1-methoxy-4-(4-methoxybenzeneseleninyl)benzene
Molecular Weight (g/mol): 309.23
Molecular Formula: C14H14O3Se
InChi Key: SXEWETDJNQVWQY-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)[Se](=O)C1=CC=C(OC)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results