accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2587-00-0

IUPAC Name: 2,6-dichloropyridin-1-ium-1-olate
Synonyms: 2,6-dichloropyridin-1-ium-1-olate
Molecular Weight (g/mol): 163.99
Molecular Formula: C5H3Cl2NO
InChi Key: RFOMGVDPYLWLOC-UHFFFAOYSA-N
SMILES: [O-][N+]1=C(Cl)C=CC=C1Cl
Percent Purity 
  • (1)
  • (5)
Quantity 
  • (3)
  • (3)
Physical Form 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (1)
  • (4)
  • (1)
  • (5)
  • (3)
  • (3)
  • (1)
  • (2)