Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dichloropyridine N-Oxide 98.0+%, TCI America™
SDP

Catalog No. D32195G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D32195G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D32195G Supplier TCI America Supplier No. D32195G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-purity building block for chemical synthesis with a minimum 98.0% purity by GC.

Chemical Identifiers

CAS 2587-00-0
Molecular Formula C5H3Cl2NO
Molecular Weight (g/mol) 163.99
MDL Number MFCD00130246
InChI Key RFOMGVDPYLWLOC-UHFFFAOYSA-N
Synonym 2,6-dichloropyridine n-oxide, 2,6-dichloropyridine 1-oxide, 2,6-dichloropyridine-n-oxide, 2,6-dichloropyridin-1-ium-1-olate, pyridine, 2,6-dichloro-, 1-oxide, 2,6-dichloropyridine-1-oxide, 2,6-dichloropyridine n-o, ide, acmc-1cjmr, 2,6-dichloropyridin-1-ol
PubChem CID 817673
IUPAC Name 2,6-dichloropyridin-1-ium-1-olate
SMILES [O-][N+]1=C(Cl)C=CC=C1Cl

Specifications

Melting Point 138°C
Color White-Yellow
Quantity 5 g
Formula Weight 163.99
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Dichloropyridine N-Oxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.