accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2587-02-2

IUPAC Name: 4-amino-2,6-dichloropyridin-1-ium
Synonyms: 4-amino-2,6-dichloropyridin-1-ium
Molecular Weight (g/mol): 164.01
Molecular Formula: C5H5Cl2N2
InChi Key: WAEZOSSWRXDWAX-UHFFFAOYSA-O
SMILES: NC1=CC(Cl)=[NH+]C(Cl)=C1
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (1)
  • (1)
  • (2)
Quantity 
  • (3)
  • (3)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (4)
  • (2)
  • (4)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)