Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dichloropyridin-4-amine, 97%, Thermo Scientific™

Catalog No. RF04500DA
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
RF04500DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RF04500DA Supplier Thermo Scientific Chemicals Supplier No. RF04500DA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2587-02-2 , 2588-04-7
Molecular Formula C5H5Cl2N2
Molecular Weight (g/mol) 164.01
InChI Key WAEZOSSWRXDWAX-UHFFFAOYSA-O
Synonym 4-amino-2,6-dichloropyridine, 2,6-dichloro-4-aminopyridine, 2,6-dichloro-pyridin-4-ylamine, 4-pyridinamine, 2,6-dichloro, 2,6-dichloro-4-pyridylamine, 2,6-dichloropyridin-4-ylamine, 2,6-dichloro-4-amino pyridine, 2,6-dichloro-4-pyridinamine, pubchem5530, acmc-1cot2
PubChem CID 137652
IUPAC Name 2,6-dichloropyridin-4-amine
SMILES C1=C(C=C(N=C1Cl)Cl)N

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 163.01
Percent Purity 97%
Chemical Name or Material 2, 6-Dichloropyridin-4-amine
Hazard Category Warning
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.