accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2587-42-0

IUPAC Name: 1,2-bis[4-(hexyloxy)phenyl]-2-oxohydrazin-2-ium-1-ide
Synonyms: 1,2-bis[4-(hexyloxy)phenyl]-2-oxohydrazin-2-ium-1-ide
Molecular Weight (g/mol): 398.55
Molecular Formula: C24H34N2O3
InChi Key: GWRSINRMEBHRIO-UHFFFAOYSA-N
SMILES: CCCCCCOC1=CC=C([N-][N+](=O)C2=CC=C(OCCCCCC)C=C2)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results